Zinc formate 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B24210.22
B24210.36
B24210.22EA
1860
CZK
B24210.22
B24210.36
Zinc formate 98%
Zinc formate
Purity: 98%
CAS No.: 557-41-5
Molecular Formula: C2H2O4Zn
Formula Weight: 155.40
UN No.: UN3077
MDL No.: MFCD00050836
Vzorec:
C₂H₂O₄Zn Storage Temperature: Ambient |
MDL Number:
MFCD00050836 CAS číslo: 557-41-5 EINECS: 209-176-5 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...