Methyl-alpha-L-rhamnopyranoside 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(2R,3R,4R,5R,6S)-2-Methoxy-6-methyltetrahydro-2H-pyran-3,4,5-triol
B24129.06
B24129.14
B24129.06EA
2250
CZK
B24129.06
B24129.14
Methyl-alpha-L-rhamnopyranoside 98%
Methyl-alpha-L-rhamnopyranoside
Purity: 98%
CAS No.: 14917-55-6
Molecular Formula: C7H14O5
Formula Weight: 178.19
Melting Point: 106-108°
MDL No.: MFCD00067654
Vzorec:
C₇H₁₄O₅ Bod tání: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067654 CAS číslo: 14917-55-6 EINECS: 238-987-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...