Tisk…

Cobalt(II) citrate dihydrate 98%

Dodavatel: Thermo Scientific
Nebezpečí

B24088.22 B24088.14
B24088.22EA 3510 CZK
B24088.22 B24088.14
Cobalt(II) citrate dihydrate 98%
Cobalt(II) citrate dihydrate

Purity: 98% CAS No.: 6424-15-3 Molecular Formula: C12H10Co3O14.2H2O Formula Weight: 591.02 (554.99anhy) Melting Point: ca 150° dec. UN No.: UN3077 MDL No.: MFCD02684007

Vzorec: C₁₂H₁₀Co₃O₁₄·2H₂O
MW: 591,02 g/mol (554,99 anhy)
Bod tání: ∼ 150 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD02684007
CAS číslo: 6424-15-3
EINECS: 212-751-3
UN: 3077
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR