Nitrofurantoin 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-[(5-Nitrofurfurylidene)amino]hydantoin
B24079.14
B24079.22
B24079.36
B24079.14EA
1510
CZK
B24079.14
B24079.22
B24079.36
Nitrofurantoin 98%
Nitrofurantoin
Purity: 98%
CAS No.: 67-20-9
Molecular Formula: C8H6N4O5
Formula Weight: 238.16
Melting Point: ca 265° dec.
MDL No.: MFCD00003224
Vzorec:
C₈H₆N₄O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00003224 CAS číslo: 67-20-9 EINECS: 200-646-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...