N-Acetyl-L-hydroxyproline 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B23767.04
B23767.09
B23767.04EA
1870
CZK
B23767.04
B23767.09
N-Acetyl-L-hydroxyproline 99%
N-Acetyl-L-hydroxyproline
Purity: 99%
CAS No.: 33996-33-7
Molecular Formula: C7H11NO4
Formula Weight: 173.17
Melting Point: 131-133°
Beilstein Reference: 84043
Optical Rotation: -117° (c=2 in water)
MDL No.: MFCD00037339
Vzorec:
C₇H₁₁NO₄ MW: 173,168 g/mol Bod tání: 131…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037339 CAS číslo: 33996-33-7 EINECS: 251-780-6 Merck Index: 14,6903 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...