Tisk…

Ethyl-2,2-diethylacetoacetate 97%

Dodavatel: Thermo Scientific

B23729.06 B23729.14
B23729.06EA 1090 CZK
B23729.06 B23729.14
Ethyl-2,2-diethylacetoacetate 97%
Ethyl-2,2-diethylacetoacetate

Purity: 97% CAS No.: 1619-57-4 Molecular Formula: C10H18O3 Formula Weight: 186.25 Refractive Index: 1.4330 Boiling Point: 84-86°/13mm Beilstein Reference: 1767891 MDL No.: MFCD00048552

Vzorec: C₁₀H₁₈O₃
MW: 186,25 g/mol
Bod varu: 84…86 °C (13 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00048552
CAS číslo: 1619-57-4
EINECS: 216-581-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR