Tisk…

Nickel(II) lactate tetrahydrate 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B23643.14 B23643.22 B23643.36
B23643.14EA 817 CZK
B23643.14 B23643.22 B23643.36
Nickel(II) lactate tetrahydrate 98%
Nickel(II) lactate tetrahydrate

Purity: 98% Molecular Formula: C6H10NiO6.4H2O Formula Weight: 308.91 (236.85anhy) UN No.: UN3077 MDL No.: MFCD02683106

Vzorec: C₆H₁₀NiO₆·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD02683106
UN: 3077
ADR: 9,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR