Tisk…

6-Acetamidohexanoic acid hydrate 99%

Dodavatel: Thermo Fisher Scientific

B23623.06 B23623.14
B23623.06EA 989 CZK
B23623.06 B23623.14
6-Acetamidohexanoic acid hydrate 99%
6-Acetamidohexanoic acid hydrate

Purity: 99% CAS No.: 57-08-9 Molecular Formula: C8H15NO3.xH2O Formula Weight: 173.21(anhy) Melting Point: 102-106° MDL No.: MFCD00004424

Vzorec: C₈H₁₅NO₃·xH₂O
MW: 173.21 g/mol (anhy)
Bod tání: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004424
CAS číslo: 57-08-9
EINECS: 200-310-8
Merck Index: 14,45

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR