Tisk…

2-(2-Aminothiazol-4-yl)acetic acid 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: (2-Amino-4-thiazolyl)acetic acid, 2-Amino-4-thiazoleacetic acid

B23534.14 B23534.22
B23534.14EA 1340 CZK
B23534.14 B23534.22
2-(2-Aminothiazol-4-yl)acetic acid 97%
2-(2-Aminothiazol-4-yl)acetic acid

Purity: 97% CAS No.: 29676-71-9 Molecular Formula: C5H6N2O2S Formula Weight: 158.18 Melting Point: ca 130° dec. Beilstein Reference: 127415 MDL No.: MFCD00010068

Vzorec: C₅H₆N₂O₂S
MW: 158,18 g/mol
Bod tání: ∼ 130 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00010068
CAS číslo: 29676-71-9
EINECS: 249-769-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR