Tisk…

S-(Carboxymethyl)-L-cysteine 97%

Dodavatel: Thermo Scientific

Synonyms: 3-[(Carboxymethyl)thio]alanine, Carbocisteine, Carbocysteine

B23487.14 B23487.22
B23487.14EA 1730 CZK
B23487.14 B23487.22
S-(Carboxymethyl)-L-cysteine 97%
S-(Carboxymethyl)-L-cysteine

Purity: 97% CAS No.: 638-23-3 Molecular Formula: C5H9NO4S Formula Weight: 179.20 Storage Temperature: 2°C to 8°C Melting Point: ca 200° dec. Beilstein Reference: 1725012 Optical Rotation: -29° (c=10 in 10% NaHCO) MDL No.: MFCD00002614

Vzorec: C₅H₉NO₄S
MW: 179,19 g/mol
Bod tání: ∼ 200 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002614
CAS číslo: 638-23-3
EINECS: 211-327-5
Merck Index: 14,1802

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR