5,7-Dimethoxycoumarin 98%
Dodavatel: Thermo Scientific
Synonyms:
Citropten, 5,7-Dimethoxy-2H-chromen-2-one
B23226.01
B23226.03
B23226.06
B23226.01EA
1340
CZK
B23226.01
B23226.03
B23226.06
5,7-Dimethoxycoumarin 98%
5,7-Dimethoxycoumarin
Purity: 98%
CAS No.: 487-06-9
Molecular Formula: C11H10O4
Formula Weight: 206.20
Melting Point: 146-149°
MDL No.: MFCD00006870
Vzorec:
C₁₁H₁₀O₄ MW: 206,20 g/mol Bod tání: 146…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006870 CAS číslo: 487-06-9 EINECS: 207-646-4 Merck Index: 14,5492 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...