Tisk…

1-Hydrazinophthalazine hydrochloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Hydralazine Hydrochloride, Hydralazine HCl, 1-Hydrazinylphthalazine hydrochloride

B22995.06 B22995.14
B22995.06EA 1450 CZK
B22995.06 B22995.14
1-Hydrazinophthalazine hydrochloride ≥98%
1-Hydrazinophthalazine hydrochloride

Purity: 98% CAS No.: 304-20-1 Molecular Formula: C8H8N4.HCl Formula Weight: 196.64 Melting Point: ca 275° dec. Beilstein Reference: 3568329 UN No.: UN2811 MDL No.: MFCD00135998

Vzorec: C₈H₈N₄
MW: 196.64 g/mol
Bod tání: ∼ 275 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00135998
CAS číslo: 304-20-1
EINECS: 206-151-0
UN: 2811
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR