Tisk…

Julolidine 98%

Dodavatel: Thermo Fisher Scientific

B22976.03 B22976.06 B22976.14
B22976.03EA 1450 CZK
B22976.03 B22976.06 B22976.14
Julolidine 98%
Julolidine

Purity: 98% CAS No.: 479-59-4 Molecular Formula: C12H15N Formula Weight: 173.26 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 32-37° Boiling Point: 170-173°/31mm Beilstein Reference: 139925 MDL No.: MFCD00006917

Vzorec: C₁₂H₁₅N
Bod varu: 170…173 °C (31 mmHg)
Bod tání: 32…37 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006917
CAS číslo: 479-59-4
EINECS: 207-535-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR