Hydroquinone-O,O'-diacetic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B22645.06
B22645.14
B22645.22
B22645.06EA
1410
CZK
B22645.06
B22645.14
B22645.22
Hydroquinone-O,O'-diacetic acid 98%
Hydroquinone-O,O'-diacetic acid
Purity: 98%
CAS No.: 2245-53-6
Molecular Formula: C10H10O6
Formula Weight: 226.19
Melting Point: ca 255° dec.
Beilstein Reference: 1984454
MDL No.: MFCD00016816
Vzorec:
C₁₀H₁₀O₆ MW: 226.19 g/mol Bod tání: ∼ 255 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00016816 CAS číslo: 2245-53-6 EINECS: 218-834-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...