1,3-Diaminoguanidine hydrochloride 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B22615.14
B22615.22
B22615.14EA
3470
CZK
B22615.14
B22615.22
1,3-Diaminoguanidine hydrochloride 98%
1,3-Diaminoguanidine hydrochloride
Purity: 98%
CAS No.: 36062-19-8
Molecular Formula: CH7N5.HCl
Formula Weight: 125.56
Melting Point: ca 181° dec.
Beilstein Reference: 3556702
MDL No.: MFCD00012948
Vzorec:
CH₇N₅ Storage Temperature: Ambient |
MDL Number:
MFCD00012948 CAS číslo: 36062-19-8 EINECS: 253-900-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...