Tisk…

Ethyl-2-bromoisovalerate 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Bromoisovaleric Acid Ethyl Ester, 2-Bromo-3-methylbutyric Acid Ethyl Ester, Ethyl 2-bromo-3-methylbutanoate, Ethyl 2-Bromo-3-methylbutyrate

B22525.06 B22525.14
B22525.06EA 735 CZK
B22525.06 B22525.14
Ethyl-2-bromoisovalerate 97%
Ethyl-2-bromoisovalerate

Purity: 97% CAS No.: 609-12-1 Molecular Formula: C7H13BrO2 Formula Weight: 209.09 Flash Point: 65°(149°F) Refractive Index: 1.4490 Odour: Pungent Boiling Point: 185-186° Beilstein Reference: 1099039 UN No.: UN3265 MDL No.: MFCD00026855

Vzorec: C₇H₁₃BrO₂
MW: 209,08 g/mol
Bod varu: 185…186 °C
Hustota: 1,276
Bod vzplanutí: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00026855
CAS číslo: 609-12-1
EINECS: 210-178-3
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR