5-Methoxy-2-methylbenzoselenazole 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B22411.03
B22411.06
B22411.03EA
1030
CZK
B22411.03
B22411.06
5-Methoxy-2-methylbenzoselenazole 97%
5-Methoxy-2-methylbenzoselenazole
Purity: 97%
CAS No.: 2946-17-0
Molecular Formula: C9H9NOSe
Formula Weight: 226.14
Flash Point:
Vzorec:
C₉H₉NOSe Bod varu: 115…117 °C (1 mmHg) Hustota: 1,490 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005558 CAS číslo: 2946-17-0 EINECS: 220-956-4 UN: 3440 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...