Tisk…

5-Methyl-2-hepten-4-one predominantly trans 99%

Dodavatel: Thermo Fisher Scientific
Varování

B22335.04 B22335.09
B22335.04EA 1680 CZK
B22335.04 B22335.09
5-Methyl-2-hepten-4-one predominantly trans 99%
5-Methyl-2-hepten-4-one

Purity: 99% CAS No.: 81925-81-7 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 55°(131°F) Refractive Index: 1.4440 Boiling Point: 169-171° UN No.: UN1224 MDL No.: MFCD00792509

Vzorec: C₈H₁₄O
Bod varu: 169…171 °C
Bod vzplanutí: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00792509
CAS číslo: 81925-81-7
UN: 1224
ADR: 3,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR