Tisk…

Ethyl-2-chloro-4,4,4-trifluoroacetoacetate 94%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate

B22270.06 B22270.14
B22270.06EA 1350 CZK
B22270.06 B22270.14
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate 94%
Ethyl-2-chloro-4,4,4-trifluoroacetoacetate

Purity: 94% CAS No.: 363-58-6 Molecular Formula: C6H6ClF3O3 Formula Weight: 218.56 Flash Point: 67°(153°F) Refractive Index: 1.3900 Boiling Point: 67-71°/35mm Beilstein Reference: 1787023 UN No.: UN1760 MDL No.: MFCD00041540

Vzorec: C₆H₆ClF₃O₃
MW: 218,556 g/mol
Bod varu: 67…71 °C (35 mmHg)
Hustota: 1,39
Bod vzplanutí: 67 °C (153 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041540
CAS číslo: 363-58-6
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR