Tisk…

Ranitidine hydrochloride 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: N-[2-[5-[(Dimethylamino)methyl]furfurylthio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine Hydrochloride, N-(2-(((5-((Dimethylamino)methyl)furan-2-yl)methyl)thio)ethyl)-N'-methyl-2-nitroethene-1,1-diamine hydrochloride

B22260.06 B22260.14
B22260.06EA 1540 CZK
B22260.06 B22260.14
Ranitidine hydrochloride 99%
Ranitidine hydrochloride

An H-2 histamine receptor antagonist.


Applications: An H-2 histamine receptor antagonist

Vzorec: C₁₃H₂₂N₄O₃S
MW: 350,86 g/mol
Bod tání: ∼ 134 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00069339
CAS číslo: 66357-59-3
EINECS: 266-333-0
Merck Index: 14,8110

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR