Tisk…

BAPTA tetrasodium salt hydrate 98%

Dodavatel: Thermo Scientific

Synonyms: BAPTA-Na4 hydrate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt hydrate, Tetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate hydrate

B22258.03 B22258.06
B22258.03EA 4950 CZK
B22258.03 B22258.06
BAPTA tetrasodium salt hydrate 98%
BAPTA tetrasodium salt hydrate

BAPTA tetrasodium salt hydrate is a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research).


BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.

Vzorec: C₂₂H₂₀N₂Na₄O₁₀·xH₂O
MW: 564,37 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD02683087
CAS číslo: 336624-09-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR