1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-(2-Chloroethyl)pyrrolidine hydrochloride
B22236.14
B22236.22
B22236.14EA
1340
CZK
B22236.14
B22236.22
1-(2-Chloroethyl)pyrrolidine hydrochloride 98%
1-(2-Chloroethyl)pyrrolidine hydrochloride
Purity: 98%
CAS No.: 7250-67-1
Molecular Formula: C6H12ClN.HCl
Formula Weight: 170.08
Melting Point: 168-173°
Beilstein Reference: 3677061
MDL No.: MFCD00012718
Vzorec:
C₆H₁₂ClN MW: 170,08 g/mol Bod tání: 168…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012718 CAS číslo: 7250-67-1 EINECS: 230-660-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...