1-(Trifluoromethyl)vinyl acetate 97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3,3,3-Trifluoroprop-1-en-2-yl acetate
B22133.18
B22133.30
B22133.18EA
13280
CZK
B22133.18
B22133.30
1-(Trifluoromethyl)vinyl acetate 97%
1-(Trifluoromethyl)vinyl acetate
Purity: 97%
CAS No.: 2247-91-8
Molecular Formula: C5H5F3O2
Formula Weight: 154.09
Storage Temperature: 2°C to 8°C
Flash Point: 2°(35°F)
Refractive Index: 1.3420
Odour: Pungent
Boiling Point: 86-88°
Beilstein Reference: 2078331
UN No.: UN3272
MDL No.: MFCD00040844
Vzorec:
C₅H₅F₃O₂ MW: 154.09 g/mol Bod varu: 86…88 °C Hustota: 1.200 Bod vzplanutí: 2 °C (35 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00040844 CAS číslo: 2247-91-8 EINECS: 218-838-2 UN: 3272 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...