Tisk…

Methyl-3-hydroxy-2-thiophenecarboxylate 97%

Dodavatel: Thermo Scientific

Synonyms: Methyl 3-hydroxythiophene-2-carboxylate

B22129.03 B22129.06 B22129.14
B22129.03EA 1040 CZK
B22129.03 B22129.06 B22129.14
Methyl-3-hydroxy-2-thiophenecarboxylate 97%
Methyl-3-hydroxy-2-thiophenecarboxylate

Purity: 97% CAS No.: 5118-06-9 Molecular Formula: C6H6O3S Formula Weight: 158.18 Melting Point: 44-48° Boiling Point: 107-109°/13mm MDL No.: MFCD00055642

Vzorec: C₆H₆O₃S
MW: 158,18 g/mol
Bod varu: 107…109 °C (13 mmHg)
Bod tání: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00055642
CAS číslo: 5118-06-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR