Tisk…
Cobalt(II) phosphate, anhydrous 98%
Dodavatel: Thermo Fisher Scientific
Varování
B22066.14 B22066.22
B22066.14EA 1380 CZK
B22066.14 B22066.22
Cobalt(II) phosphate, anhydrous 98%
Cobalt(II) phosphate
Purity: 98% CAS No.: 13455-36-2 Molecular Formula: Co3O8P2 Formula Weight: 366.74 MDL No.: MFCD00016028
Vzorec: Co₃O₈P₂
MW: 366.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016028
CAS číslo: 13455-36-2
EINECS: 236-655-6