Tisk…

Mesitylenesulphonic acid sodium salt hemihydrate 98%

Dodavatel: Thermo Fisher Scientific

B22029.14 B22029.22 B22029.36
B22029.14EA 1530 CZK
B22029.14 B22029.22 B22029.36
Mesitylenesulphonic acid sodium salt hemihydrate 98%
Mesitylenesulphonic acid sodium salt hemihydrate

Purity: 98% CAS No.: 6148-75-0 Molecular Formula: C9H11NaO3S.0.5H2O Formula Weight: 231.26 (222.25anhy) Melting Point: 300° Beilstein Reference: 5784990 MDL No.: MFCD00067712

Vzorec: C₉H₁₁NaO₃S·¹/₂H₂O
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00067712
CAS číslo: 6148-75-0
EINECS: 228-157-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR