BAPTA tetraethyl ester ≥98%
Dodavatel: Thermo Fisher Scientific
B21971.03
B21971.06
B21971.03EA
2950
CZK
B21971.03
B21971.06
BAPTA tetraethyl ester ≥98%
BAPTA tetraethyl ester
Purity: 98+%
CAS No.: 73630-07-6
Molecular Formula: C30H40N2O10
Formula Weight: 588.66
Melting Point: 98-102°
MDL No.: MFCD00532663
Vzorec:
C₃₀H₄₀N₂O₁₀ Bod tání: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00532663 CAS číslo: 73630-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...