Tisk…

Sodium ethanesulphonate, (max. 2% H₂O) 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethanesulphonic acid Sodium Salt, Ethylsulphonic acid Sodium Salt

B21824.06 B21824.14
B21824.06EA 2310 CZK
B21824.06 B21824.14
Sodium ethanesulphonate, (max. 2% H₂O) 98%
Sodium ethanesulphonate

Purity: 98% CAS No.: 5324-47-0 Molecular Formula: CH3CH2SO3Na Formula Weight: 132.11 Melting Point: ca 270° MDL No.: MFCD00066498

Vzorec: C₂H₆O₃S
MW: 132,11 g/mol
Bod tání: ∼ 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00066498
CAS číslo: 5324-47-0
EINECS: 226-194-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR