Tisk…

9-(Trimethylsilyl)fluorene 99%

Dodavatel: Thermo Fisher Scientific

B21816.03 B21816.06
B21816.03EA 4090 CZK
B21816.03 B21816.06
9-(Trimethylsilyl)fluorene 99%
9-(Trimethylsilyl)fluorene

Purity: 99% CAS No.: 7385-10-6 Molecular Formula: C16H18Si Formula Weight: 238.40 Melting Point: 95-98° MDL No.: MFCD00092570

Vzorec: C₁₆H₁₈Si
MW: 238.40 g/mol
Bod tání: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00092570
CAS číslo: 7385-10-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR