Croconic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4,5-Dihydroxy-4-cyclopentene-1,2,3-trione, 4,5-Dihydroxycyclopent-4-ene-1,2,3-trione
B21809.03
B21809.06
B21809.03EA
5290
CZK
B21809.03
B21809.06
Croconic acid 98%
Croconic acid
Purity: 98%
CAS No.: 488-86-8
Molecular Formula: C5H2O5
Formula Weight: 142.07
Odour: Odourless
Melting Point: ca 280° dec.
MDL No.: MFCD00181389
Vzorec:
C₅H₂O₅ MW: 142,07 g/mol Bod tání: ∼ 280 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00181389 CAS číslo: 488-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...