Naphthalene-1-sulphonic acid hydrate 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B21782.09
B21782.18
B21782.09EA
1610
CZK
B21782.09
B21782.18
Naphthalene-1-sulphonic acid hydrate 98%
Naphthalene-1-sulphonic acid hydrate
Purity: 98%
CAS No.: 6036-48-2
Molecular Formula: C10H8O3S.xH2O
Formula Weight: 208.23(anhy)
Melting Point: 89-91°(dried)
UN No.: UN2585
MDL No.: MFCD00003987
Vzorec:
C₁₀H₈O₃S·xH₂O MW: 208,23 g/mol (anhy) Bod tání: 89…91 °C (dried) Storage Temperature: Ambient |
MDL Number:
MFCD00003987 CAS číslo: 6036-48-2 EINECS: 201-610-1 UN: 2585 ADR: 8,III Merck Index: 14,6376 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...