1-(1,3-Thiazol-2-yl)propan-1-one 99%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
2-Propionylthiazole, 1-(Thiazol-2-yl)propan-1-one
B21711.04
B21711.09
B21711.04EA
1370
CZK
B21711.04
B21711.09
1-(1,3-Thiazol-2-yl)propan-1-one 99%
1-(1,3-Thiazol-2-yl)propan-1-one
Purity: 99%
CAS No.: 43039-98-1
Molecular Formula: C6H7NOS
Formula Weight: 141.19
Flash Point:
Vzorec:
C₆H₇NOS MW: 141,19 g/mol Bod varu: 110 °C (5 mmHg) Hustota: 1,174 Bod vzplanutí: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01681404 CAS číslo: 43039-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...