Tisk…

N-Carbobenzoxy-L-proline ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline

B21672.06 B21672.18 B21672.30
B21672.06EA 1210 CZK
B21672.06 B21672.18 B21672.30
N-Carbobenzoxy-L-proline ≥98%
N-Carbobenzoxy-L-proline

Purity: 98+% CAS No.: 1148-11-4 Molecular Formula: C13H15NO4 Formula Weight: 249.27 Melting Point: 75-77° Beilstein Reference: 88579 Optical Rotation: -60° (c=1 in acetic acid) MDL No.: MFCD00003170

Vzorec: C₁₃H₁₅NO₄
MW: 249.27 g/mol
Bod tání: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00003170
CAS číslo: 1148-11-4
EINECS: 214-557-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR