Glibenclamide 99%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulphamoyl)phenethyl)-2-methoxybenzamide, 5-Chloro-N-[4-(3-cyclohexylureidosulphonyl)phenethyl]-2-methoxybenzamide, Glyburide
B21459.14
B21459.22
B21459.14EA
3090
CZK
B21459.14
B21459.22
Glibenclamide 99%
Glibenclamide
Purity: 99%
CAS No.: 10238-21-8
Molecular Formula: C23H28ClN3O5S
Formula Weight: 494.01
Storage Temperature: 2°C to 8°C
Melting Point: 171-174°
MDL No.: MFCD00056625
Vzorec:
C₂₃H₂₈ClN₃O₅S MW: 494,00 g/mol Bod tání: 171…174 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00056625 CAS číslo: 10238-21-8 EINECS: 233-570-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...