Tisk…

4,5,6-Triaminopyrimidine sulphate hydrate, (max. 8% H₂O) ≥98% (dry weight)

Dodavatel: Thermo Fisher Scientific
Varování

B21369.03 B21369.06
B21369.03EA 1940 CZK
B21369.03 B21369.06
4,5,6-Triaminopyrimidine sulphate hydrate, (max. 8% H₂O) ≥98% (dry weight)
4,5,6-Triaminopyrimidine sulphate hydrate

Purity: 98+% CAS No.: 207742-76-5 Molecular Formula: C4H7N5.H2SO4.xH2O Formula Weight: 223.22(anhy) Melting Point: 300° MDL No.: MFCD00012789

Vzorec: C₄H₇N₅·H₂SO₄·xH₂O
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012789
CAS číslo: 207742-76-5
EINECS: 256-446-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR