Tisk…

3-Chloropropyl phenyl sulphide 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B21345.06 B21345.14
B21345.06EA 1860 CZK
B21345.06 B21345.14
3-Chloropropyl phenyl sulphide 97%
3-Chloropropyl phenyl sulphide

Purity: 97% CAS No.: 4911-65-3 Molecular Formula: C9H11ClS Formula Weight: 186.70 Odour: Stench MDL No.: MFCD00040122

Vzorec: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD00040122
CAS číslo: 4911-65-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR