Tisk…

Diethyl (phthalimidomethyl)phosphonate 97%

Dodavatel: Thermo Fisher Scientific

Synonyms: Diethyl ((1,3-dioxoisoindolin-2-yl)methyl)phosphonate, (Phthalimidomethyl)phosphonic Acid Diethyl Ester

B21096.06 B21096.14
B21096.06EA 2030 CZK
B21096.06 B21096.14
Diethyl (phthalimidomethyl)phosphonate 97%
Diethyl (phthalimidomethyl)phosphonate

Purity: 97% CAS No.: 33512-26-4 Molecular Formula: C13H16NO5P Formula Weight: 297.25 Melting Point: 60-64° MDL No.: MFCD00134313

Vzorec: C₁₃H₁₆NO₅P
Bod tání: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00134313
CAS číslo: 33512-26-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR