N-Fmoc-L-serine ≥97%
Dodavatel: Thermo Scientific
Synonyms:
Fmoc-Ser-OH
B21079.03
B21079.06
B21079.03EA
999
CZK
B21079.03
B21079.06
N-Fmoc-L-serine ≥97%
N-Fmoc-L-serine
Purity: 97+%
CAS No.: 73724-45-5
Molecular Formula: C18H17NO5
Formula Weight: 327.34
Storage Temperature: 2°C to 8°C
Melting Point: 102-106°
Beilstein Reference: 4715791
Optical Rotation: +13.6° (c=1 in ethyl acetate)
MDL No.: MFCD00051928
Vzorec:
C₁₈H₁₇NO₅ MW: 327,34 g/mol Bod tání: 102…106 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051928 CAS číslo: 73724-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...