4-Methoxy-4-methyl-2-pentanone 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B21076.14
B21076.22
B21076.14EA
1500
CZK
B21076.14
B21076.22
4-Methoxy-4-methyl-2-pentanone 97%
4-Methoxy-4-methyl-2-pentanone
Purity: 97%
CAS No.: 107-70-0
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 48°(118°F)
Refractive Index: 1.4180
Odour: Mild
Boiling Point: 156°
UN No.: UN2293
MDL No.: MFCD00043932
Vzorec:
C₇H₁₄O₂ Bod varu: 156 °C Hustota: 0,91 Bod vzplanutí: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043932 CAS číslo: 107-70-0 EINECS: 203-512-4 UN: 2293 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...