Tisk…

6-Hydroxy-2-methylpyridazin-3(2H)-one 97%

Dodavatel: Thermo Scientific

B20975.06 B20975.14
B20975.06EA 2030 CZK
B20975.06 B20975.14
6-Hydroxy-2-methylpyridazin-3(2H)-one 97%
6-Hydroxy-2-methylpyridazin-3(2H)-one

Purity: 97% CAS No.: 5436-01-1 Molecular Formula: C5H6N2O2 Formula Weight: 126.11 Melting Point: 216-219° MDL No.: MFCD00234108

Vzorec: C₅H₆N₂O₂
MW: 126,12 g/mol
Bod tání: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00176268
CAS číslo: 5436-01-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR