Tisk…
2-Methyl-1-phenyl-2-propen-1-ol ≥85%, tech.
Dodavatel: Thermo Fisher Scientific
B20936.06 B20936.14
B20936.06EA 1810 CZK
B20936.06 B20936.14
2-Methyl-1-phenyl-2-propen-1-ol ≥85%, tech.
2-Methyl-1-phenyl-2-propen-1-ol
Purity: 85% CAS No.: 4383-08-8 Molecular Formula: C10H12O Formula Weight: 148.21 MDL No.: MFCD00046630
Vzorec: C₁₀H₁₂O
MW: 148.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046630
CAS číslo: 4383-08-8