Tisk…

2,4-Diaminodiphenylamine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: N1-Phenylbenzene-1,2,4-triamine

B20735.03 B20735.06
B20735.03EA 1240 CZK
B20735.03 B20735.06
2,4-Diaminodiphenylamine 98%
2,4-Diaminodiphenylamine

Purity: 98% CAS No.: 136-17-4 Molecular Formula: C12H13N3 Formula Weight: 199.26 Melting Point: 129-131° UN No.: UN2811 MDL No.: MFCD00025285

Vzorec: C₁₂H₁₃N₃
MW: 199,26 g/mol
Bod tání: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00025285
CAS číslo: 136-17-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR