Tisk…

N,N'-Bis(2-hydroxyethyl)oxamide 99%

Dodavatel: Thermo Scientific

Synonyms: N,N'-Oxalylbis(ethanolamine), N1,N2-Bis(2-hydroxyethyl)oxalamide

B20458.06 B20458.14 B20458.22
B20458.06EA 1560 CZK
B20458.06 B20458.14 B20458.22
N,N'-Bis(2-hydroxyethyl)oxamide 99%
N,N'-Bis(2-hydroxyethyl)oxamide

Purity: 99% CAS No.: 1871-89-2 Molecular Formula: C6H12N2O4 Formula Weight: 176.17 Melting Point: 167-169° dec. MDL No.: MFCD00020559

Vzorec: C₆H₁₂N₂O₄
MW: 176,17 g/mol
Bod tání: 167…169 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00020559
CAS číslo: 1871-89-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR