Tisk…
Di-n-propyl succinate 98%
Dodavatel: Thermo Fisher Scientific
B20441.09 B20441.18
B20441.09EA 769 CZK
B20441.09 B20441.18
Di-n-propyl succinate 98%
Di-n-propyl succinate
Purity: 98% CAS No.: 925-15-5 Molecular Formula: C10H18O4 Formula Weight: 202.25 Odour: Pleasant Boiling Point: 250-252° MDL No.: MFCD00015213
Vzorec: C₁₀H₁₈O₄
MW: 202.25 g/mol
Bod varu: 250…252 °C
Hustota: 1.00
Storage Temperature: Ambient
MDL Number: MFCD00015213
CAS číslo: 925-15-5
EINECS: 213-114-2