Cyclopentyl phenyl ketone 96%
Dodavatel: Thermo Scientific
Synonyms:
Cyclopentyl(phenyl)methanone, Benzoylcyclopentane
A19976.03
A19976.06
A19976.03EA
882
CZK
A19976.03
A19976.06
Cyclopentyl phenyl ketone 96%
Cyclopentyl phenyl ketone
Purity: 96%
CAS No.: 5422-88-8
Molecular Formula: C12H14O
Formula Weight: 174.24
Flash Point:
Vzorec:
C₁₂H₁₄O MW: 174,24 g/mol Bod varu: 136…140 °C (16 mmHg) Hustota: 1,036 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001378 CAS číslo: 5422-88-8 EINECS: 226-548-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...