Tisk…

Brucine sulphate hydrate, (∼13% H₂O) 98% (dry weight)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A19943.22 A19943.14
A19943.22EA 3750 CZK
A19943.22 A19943.14
Brucine sulphate hydrate, (∼13% H₂O) 98% (dry weight)
Brucine sulphate hydrate

Purity: 98% CAS No.: 652154-10-4 Molecular Formula: C23H26N2O4.0.5H2SO4.xH2O Formula Weight: 443.52(anhy) Melting Point: 191-196° dec. UN No.: UN2811 MDL No.: MFCD00013472

Vzorec: C₂₃H₂₆N₂O₄·¹/₂H₂SO₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00013472
CAS číslo: 652154-10-4
EINECS: 225-432-9
UN: 2811
ADR: 6.1,I
Merck Index: 14,1455

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR