Tisk…

Sodium-3-bromopropanesulphonate 97%

Dodavatel: Thermo Fisher Scientific
Varování

A19930.06 A19930.22
A19930.06EA 1110 CZK
A19930.06 A19930.22
Sodium-3-bromopropanesulphonate 97%
Sodium-3-bromopropanesulphonate

Purity: 97% CAS No.: 55788-44-8 Molecular Formula: Br(CH2)3SO3Na Formula Weight: 225.05 Melting Point: ca 256° dec. MDL No.: MFCD00024999

Vzorec: C₃H₇BrO₃S
Storage Temperature: Ambient
MDL Number: MFCD00024999
CAS číslo: 55788-44-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR