Tisk…

1,2-Bis(methanesulphonamido)benzene 97%

Dodavatel: Thermo Scientific

A19830.09 A19830.88
A19830.09EA 3400 CZK
A19830.09 A19830.88
1,2-Bis(methanesulphonamido)benzene 97%
1,2-Bis(methanesulphonamido)benzene

Purity: 97% CAS No.: 7596-80-7 Molecular Formula: C8H12N2O4S2 Formula Weight: 264.32 Odour: Odourless Melting Point: 211-214° MDL No.: MFCD00276605

Vzorec: C₈H₁₂N₂O₄S₂
MW: 264,32 g/mol
Bod tání: 211…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00276605
CAS číslo: 7596-80-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR