5,6,7,8,9,10-Hexahydrocyclohept[b]indole 98%
Dodavatel: Thermo Fisher Scientific
A19344.03
A19344.06
A19344.03EA
2640
CZK
A19344.03
A19344.06
5,6,7,8,9,10-Hexahydrocyclohept[b]indole 98%
5,6,7,8,9,10-Hexahydrocyclohept[b]indole
Purity: 98%
CAS No.: 2047-89-4
Molecular Formula: C13H15N
Formula Weight: 185.27
Melting Point: 143-146°
MDL No.: MFCD00101340
Vzorec:
C₁₃H₁₅N Bod tání: 143…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00101340 CAS číslo: 2047-89-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...