Pyridine hydrobromide 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A19314.0B
A19314.18
A19314.30
A19314.0BEA
6970
CZK
A19314.0B
A19314.18
A19314.30
Pyridine hydrobromide 98%
Pyridine hydrobromide
Purity: 98%
CAS No.: 18820-82-1
Molecular Formula: C5H5N.HBr
Formula Weight: 160.01
Melting Point: 216-220°
MDL No.: MFCD00012801
Vzorec:
C₅H₅N Bod tání: 216…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012801 CAS číslo: 18820-82-1 EINECS: 242-600-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...